Tisk…

tert-Butyl 4-[2-[methoxy(methyl)amino]-2-oxoethyl]piperidine-1-carboxylate 96%

Dodavatel: Apollo Scientific

OR952997-1G OR952997-250MG
APOSOR952997-1GEA 8530 CZK
APOSOR952997-1G APOSOR952997-250MG
tert-Butyl 4-[2-[methoxy(methyl)amino]-2-oxoethyl]piperidine-1-carboxylate 96%
tert-Butyl 4-[2-[methoxy(methyl)amino]-2-oxoethyl]piperidine-1-carboxylate
Vzorec: C₁₄H₂₆N₂O₄
MW: 286,367 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07370046
CAS číslo: 416852-69-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR