Tisk…

Isoquinoline-4-carbothioic acid amide 95%

Dodavatel: Apollo Scientific

OR952895-1G OR952895-250MG
APOSOR952895-1GEA 6910 CZK
APOSOR952895-1G APOSOR952895-250MG
Isoquinoline-4-carbothioic acid amide 95%
Isoquinoline-4-carbothioic acid amide
Vzorec: C₁₀H₈N₂S
Storage Temperature: Ambient
MDL Number: MFCD06738908
CAS číslo: 435271-32-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR