Tisk…

N-(4-Ethylbenzyl)-N-methylamine 95%

Dodavatel: Apollo Scientific

Synonyms: 1-(4-Ethylphenyl)-N-methylmethanamine

OR952845-1G OR952845-5G
APOSOR952845-1GEA 2110 CZK
APOSOR952845-1G APOSOR952845-5G
N-(4-Ethylbenzyl)-N-methylamine 95%
N-(4-Ethylbenzyl)-N-methylamine
Vzorec: C₁₀H₁₅N
MW: 149,233 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04115409
CAS číslo: 568577-84-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR