Tisk…

Azepan-3-yl-methyl-amine 95%

Dodavatel: Apollo Scientific

OR952842-1G OR952842-250MG
APOSOR952842-1GEA 17990 CZK
APOSOR952842-1G APOSOR952842-250MG
Azepan-3-yl-methyl-amine 95%
Azepan-3-yl-methyl-amine
Vzorec: C₇H₁₆N₂
MW: 128,215 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02683066
CAS číslo: 124695-93-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR