Tisk…

(4-tert-Butoxy-phenyl)-methanol 95%

Dodavatel: Apollo Scientific

Synonyms: 4-tert-Butoxybenzyl Alcohol

OR952840-1G OR952840-250MG OR952840-5G
APOSOR952840-1GEA 8070 CZK
APOSOR952840-1G APOSOR952840-250MG APOSOR952840-5G
(4-tert-Butoxy-phenyl)-methanol 95%
(4-tert-Butoxy-phenyl)-methanol
Vzorec: C₁₁H₁₆O₂
MW: 180,244 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00082599
CAS číslo: 51503-08-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR