4-(Methoxymethyl)aniline 96%
Dodavatel: Apollo Scientific
OR952839-10G
OR952839-1G
OR952839-5G
APOSOR952839-10GEA
5720
CZK
APOSOR952839-10G
APOSOR952839-1G
APOSOR952839-5G
4-(Methoxymethyl)aniline 96%
4-(Methoxymethyl)aniline
Vzorec:
C₈H₁₁NO |
MDL Number:
MFCD06804487 CAS číslo: 80936-82-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...