Tisk…

8-Chloro-2,3,4,5-tetrahydro-1H-benzo[e][1,4]diazepine 95%

Dodavatel: Apollo Scientific

OR952723-25G OR952723-5G
APOSOR952723-25GEA 14290 CZK
APOSOR952723-25G APOSOR952723-5G
8-Chloro-2,3,4,5-tetrahydro-1H-benzo[e][1,4]diazepine 95%
8-Chloro-2,3,4,5-tetrahydro-1H-benzo[e][1,4]diazepine
Vzorec: C₉H₁₁ClN₂
MW: 182,65 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07369929
CAS číslo: 107479-55-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR