(2-Formyl-4-nitrophenoxy)acetic acid
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-(2-Formyl-4-nitrophenoxy)ethanoic acid, 2-(2-Formyl-4-nitrophenoxy)acetic acid, 2-(Carboxymethoxy)-5-nitrobenzaldehyde
OR9527-1G
OR9527-5G
OR9527-25G
APOSOR9527-1GEA
622
CZK
APOSOR9527-1G
APOSOR9527-5G
APOSOR9527-25G
(2-Formyl-4-nitrophenoxy)acetic acid
(2-Formyl-4-nitrophenoxy)acetic acid
Vzorec:
C₉H₇NO₆ MW: 225,155 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00024502 CAS číslo: 6965-69-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...