Tisk…

4-(1-Methyl-5-imidazolyl)aniline 95%

Dodavatel: Apollo Scientific

OR952608-100MG OR952608-1G OR952608-250MG
APOSOR952608-100MGEA 4140 CZK
APOSOR952608-100MG APOSOR952608-1G APOSOR952608-250MG
4-(1-Methyl-5-imidazolyl)aniline 95%
4-(1-Methyl-5-imidazolyl)aniline
Vzorec: C₁₀H₁₁N₃
MW: 173,214 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18375273
CAS číslo: 89250-15-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR