7-Chloro-3,4-dihydro-1H-quinoxalin-2-one 95%
Dodavatel: Apollo Scientific
OR952591-1G
OR952591-5G
APOSOR952591-1GEA
10250
CZK
APOSOR952591-1G
APOSOR952591-5G
7-Chloro-3,4-dihydro-1H-quinoxalin-2-one 95%
7-Chloro-3,4-dihydro-1H-quinoxalin-2-one
Vzorec:
C₈H₇ClN₂O MW: 182,607 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03426380 CAS číslo: 66367-05-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...