Tisk…

2-Amino-3-cyclopropylpyridine

Dodavatel: Apollo Scientific
Varování

Synonyms: 3-Cyclopropylpyridin-2-amine

OR952526-1G OR952526-250MG OR952526-5G
APOSOR952526-1GEA 8240 CZK
APOSOR952526-1G APOSOR952526-250MG APOSOR952526-5G
2-Amino-3-cyclopropylpyridine
2-Amino-3-cyclopropylpyridine
Vzorec: C₈H₁₀N₂
Storage Temperature: Refrigerator
MDL Number: MFCD13175917
CAS číslo: 878805-25-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR