Tisk…

4-tert-Butyl 3-methyl morpholine-3,4-dicarboxylate 97%

Dodavatel: Apollo Scientific
Varování

OR952491-1G OR952491-250MG OR952491-5G
APOSOR952491-1GEA 15740 CZK
APOSOR952491-1G APOSOR952491-250MG APOSOR952491-5G
4-tert-Butyl 3-methyl morpholine-3,4-dicarboxylate 97%
4-tert-Butyl 3-methyl morpholine-3,4-dicarboxylate
Vzorec: C₁₁H₁₉NO₅
MW: 245,272 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06656782
CAS číslo: 212650-45-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR