Tisk…

N-Ethyl-3,4-dimethylbenzylamine 95%

Dodavatel: Apollo Scientific

OR952456-1G OR952456-5G
APOSOR952456-1GEA 6420 CZK
APOSOR952456-1G APOSOR952456-5G
N-Ethyl-3,4-dimethylbenzylamine 95%
N-Ethyl-3,4-dimethylbenzylamine
Vzorec: C₁₁H₁₇N
MW: 163,259 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11204729
CAS číslo: 39180-83-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR