Tisk…

6-Methoxyisoquinolin-1(2H)-one 95%

Dodavatel: Apollo Scientific

OR952438-1G
APOSOR952438-1GEA 4070 CZK
APOSOR952438-1G
6-Methoxyisoquinolin-1(2H)-one 95%
6-Methoxyisoquinolin-1(2H)-one
Vzorec: C₁₀H₉NO₂
MW: 175,184 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12405870
CAS číslo: 26829-43-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR