6-Cyclohexyl-3(2H)-pyridazinone 95%
Dodavatel: Apollo Scientific
OR952326-1G
OR952326-250MG
APOSOR952326-1GEA
21490
CZK
APOSOR952326-1G
APOSOR952326-250MG
6-Cyclohexyl-3(2H)-pyridazinone 95%
6-Cyclohexyl-3(2H)-pyridazinone
Vzorec:
C₁₀H₁₄N₂O MW: 178,231 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11520598 CAS číslo: 105537-90-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...