2,6-Diamino-3,5-dinitropyridine
Dodavatel: Apollo Scientific
|
Varování
|
OR9523-1G
OR9523-5G
OR9523-25G
APOSOR9523-1GEA
2480
CZK
APOSOR9523-1G
APOSOR9523-5G
APOSOR9523-25G
2,6-Diamino-3,5-dinitropyridine
2,6-Diamino-3,5-dinitropyridine
Vzorec:
C₅H₅N₅O₄ MW: 199.124 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00224331 CAS číslo: 34981-11-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...