Tisk…

3-(Methoxymethyl)aniline 98%

Dodavatel: Apollo Scientific

OR952259-10G OR952259-1G OR952259-5G
APOSOR952259-10GEA 20470 CZK
APOSOR952259-10G APOSOR952259-1G APOSOR952259-5G
3-(Methoxymethyl)aniline 98%
3-(Methoxymethyl)aniline
Vzorec: C₈H₁₁NO
MW: 137,179 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06804494
CAS číslo: 53473-83-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR