Tisk…

4-Methoxybutanoic acid 95%

Dodavatel: Apollo Scientific

OR952214-1G OR952214-250MG
APOSOR952214-1GEA 19840 CZK
APOSOR952214-1G APOSOR952214-250MG
4-Methoxybutanoic acid 95%
4-Methoxybutanoic acid
Vzorec: C₅H₁₀O₃
MW: 118,131 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00029855
CAS číslo: 29006-02-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR