Tisk…

Ethyl 3-hydroxypyridazine-4-carboxylate 95%

Dodavatel: Apollo Scientific

OR952193-1G OR952193-250MG
APOSOR952193-1GEA 7770 CZK
APOSOR952193-1G APOSOR952193-250MG
Ethyl 3-hydroxypyridazine-4-carboxylate 95%
Ethyl 3-hydroxypyridazine-4-carboxylate
Vzorec: C₇H₈N₂O₃
MW: 168,15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12828686
CAS číslo: 1445-55-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR