Tisk…

2-Chloro-6-methylbenzoyl chloride 96%

Dodavatel: Apollo Scientific

OR952170-1G OR952170-5G
APOSOR952170-1GEA 3060 CZK
APOSOR952170-1G APOSOR952170-5G
2-Chloro-6-methylbenzoyl chloride 96%
2-Chloro-6-methylbenzoyl chloride
Vzorec: C₈H₆Cl₂O
MW: 189,039 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12025218
CAS číslo: 89894-44-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR