Tisk…

Methyl[2-(3-methylphenoxy)ethyl]amine 96%

Dodavatel: Apollo Scientific

OR952079-1G OR952079-5G
APOSOR952079-1GEA 4770 CZK
APOSOR952079-1G APOSOR952079-5G
Methyl[2-(3-methylphenoxy)ethyl]amine 96%
Methyl[2-(3-methylphenoxy)ethyl]amine
Vzorec: C₁₀H₁₅NO
MW: 165,232 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00085971
CAS číslo: 200350-18-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR