Tisk…

Methyl (4-cyanophenoxy)acetate 98%

Dodavatel: Apollo Scientific

OR952046-1G OR952046-25G OR952046-5G
APOSOR952046-1GEA 695 CZK
APOSOR952046-1G APOSOR952046-25G APOSOR952046-5G
Methyl (4-cyanophenoxy)acetate 98%
Methyl (4-cyanophenoxy)acetate
Vzorec: C₁₀H₉NO₃
Storage Temperature: Ambient
MDL Number: MFCD05855148
CAS číslo: 272792-14-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR