Tisk…
6,7-Dihydro-5H-[1,3]dioxolo[4,5-f]isoindole hydrochloride 95%
Dodavatel: Apollo Scientific
OR951958-1G OR951958-250MG
APOSOR951958-1GEA 32930 CZK
APOSOR951958-1G APOSOR951958-250MG
6,7-Dihydro-5H-[1,3]dioxolo[4,5-f]isoindole hydrochloride 95%
6,7-Dihydro-5H-[1,3]dioxolo[4,5-f]isoindole hydrochloride
Vzorec: C₉H₉NO₂·ClH
MW: 199,634 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17926273
CAS číslo: 1255099-33-2