Tisk…

5-Methyl-2-phenyl-1H-imidazole-4-methanol 97%

Dodavatel: Apollo Scientific

Synonyms: 4-Hydroxymethyl-5-methyl-2-phenylimidazole

OR951906-1G
APOSOR951906-1GEA 2110 CZK
APOSOR951906-1G
5-Methyl-2-phenyl-1H-imidazole-4-methanol 97%
5-Methyl-2-phenyl-1H-imidazole-4-methanol
Vzorec: C₁₁H₁₂N₂O
MW: 188,226 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00047016
CAS číslo: 13682-32-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR