N-(1-Phenyl-ethyl)-acetamide 95%
Dodavatel: Apollo Scientific
OR951896-1G
OR951896-25G
OR951896-5G
APOSOR951896-1GEA
744
CZK
APOSOR951896-1G
APOSOR951896-25G
APOSOR951896-5G
N-(1-Phenyl-ethyl)-acetamide 95%
N-(1-Phenyl-ethyl)-acetamide
Vzorec:
C₁₀H₁₃NO Storage Temperature: Ambient |
MDL Number:
MFCD00040690 CAS číslo: 36065-27-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...