Tisk…

b-Amino-4-methoxybenzenepropanol 97%

Dodavatel: Apollo Scientific

Synonyms: 2-Amino-3-(4-methoxyphenyl)propan-1-ol

OR951852-1G OR951852-250MG
APOSOR951852-1GEA 24730 CZK
APOSOR951852-1G APOSOR951852-250MG
b-Amino-4-methoxybenzenepropanol 97%
b-Amino-4-methoxybenzenepropanol
Vzorec: C₁₀H₁₅NO₂
MW: 181,232 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10034617
CAS číslo: 176035-15-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR