Tisk…

5-Chloro-1,2,3,4-tetrahydroisoquinoline 95%

Dodavatel: Apollo Scientific
Varování

OR951833-1G OR951833-250MG
APOSOR951833-1GEA 16140 CZK
APOSOR951833-1G APOSOR951833-250MG
5-Chloro-1,2,3,4-tetrahydroisoquinoline 95%
5-Chloro-1,2,3,4-tetrahydroisoquinoline
Vzorec: C₉H₁₀ClN
MW: 167,635 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08544277
CAS číslo: 73075-43-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR