5-Formyl-2-methoxybenzoic acid 96%
Dodavatel: Apollo Scientific
OR951686-25G
OR951686-5G
APOSOR951686-25GEA
16800
CZK
APOSOR951686-25G
APOSOR951686-5G
5-Formyl-2-methoxybenzoic acid 96%
5-Formyl-2-methoxybenzoic acid
Vzorec:
C₉H₈O₄ Storage Temperature: Ambient |
MDL Number:
MFCD01442588 CAS číslo: 84923-70-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...