5-Hydroxy-2-(methylthio)pyrimidine 95%
Dodavatel: Apollo Scientific
Synonyms:
2-(Methylthio)pyrimidin-5-ol
OR951682-1G
OR951682-250MG
APOSOR951682-1GEA
7740
CZK
APOSOR951682-1G
APOSOR951682-250MG
5-Hydroxy-2-(methylthio)pyrimidine 95%
5-Hydroxy-2-(methylthio)pyrimidine
Vzorec:
C₅H₆N₂OS MW: 142,179 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00223737 CAS číslo: 4874-33-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...