Tisk…

4-(Pyrrolidin-1-ylmethyl)-1,3-thiazol-2-amine 95%

Dodavatel: Apollo Scientific

OR951675-100MG OR951675-1G OR951675-250MG
APOSOR951675-100MGEA 4140 CZK
APOSOR951675-100MG APOSOR951675-1G APOSOR951675-250MG
4-(Pyrrolidin-1-ylmethyl)-1,3-thiazol-2-amine 95%
4-(Pyrrolidin-1-ylmethyl)-1,3-thiazol-2-amine
Vzorec: C₈H₁₃N₃S
MW: 183,274 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00490015
CAS číslo: 17386-09-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR