Tisk…

5-(Methylamino)nicotinic acid 95%

Dodavatel: Apollo Scientific

OR951641-1G OR951641-250MG
APOSOR951641-1GEA 8870 CZK
APOSOR951641-1G APOSOR951641-250MG
5-(Methylamino)nicotinic acid 95%
5-(Methylamino)nicotinic acid
Vzorec: C₇H₈N₂O₂
MW: 152,151 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04971939
CAS číslo: 91702-88-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR