Tisk…
Methyl 1,3-benzodioxole-4-carboxylate 98%
Dodavatel: Apollo Scientific
Synonyms: Methyl benzo[d][1,3]dioxole-4-carboxylate
OR951639-1G OR951639-5G
APOSOR951639-1GEA 827 CZK
APOSOR951639-1G APOSOR951639-5G
Methyl 1,3-benzodioxole-4-carboxylate 98%
Methyl 1,3-benzodioxole-4-carboxylate
Vzorec: C₉H₈O₄
MW: 180,157 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01546796
CAS číslo: 33842-16-9