N-Ethyl-(2-thienylmethyl)amine 95%
Dodavatel: Apollo Scientific
OR951624-1G
OR951624-5G
APOSOR951624-1GEA
4140
CZK
APOSOR951624-1G
APOSOR951624-5G
N-Ethyl-(2-thienylmethyl)amine 95%
N-Ethyl-(2-thienylmethyl)amine
Vzorec:
C₇H₁₁NS Storage Temperature: Ambient |
MDL Number:
MFCD04478205 CAS číslo: 58255-25-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...