1-Phenyl-1H-imidazole-5-carboxylic acid 97%
Dodavatel: Apollo Scientific
OR951600-1G
OR951600-250MG
APOSOR951600-1GEA
2730
CZK
APOSOR951600-1G
APOSOR951600-250MG
1-Phenyl-1H-imidazole-5-carboxylic acid 97%
1-Phenyl-1H-imidazole-5-carboxylic acid
Vzorec:
C₁₀H₈N₂O₂ MW: 188,183 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06655358 CAS číslo: 135417-65-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...