Tisk…

2-(Methylsulphonyl)benzylamine 98%

Dodavatel: Apollo Scientific

OR951580-1G OR951580-5G
APOSOR951580-1GEA 7050 CZK
APOSOR951580-1G APOSOR951580-5G
2-(Methylsulphonyl)benzylamine 98%
2-(Methylsulphonyl)benzylamine
Vzorec: C₈H₁₁NO₂S
Storage Temperature: Ambient
MDL Number: MFCD08234913
CAS číslo: 410545-65-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR