5-Amino-1-ethyl-1H-indazole 98%
Dodavatel: Apollo Scientific
OR951560-1G
APOSOR951560-1GEA
3440
CZK
APOSOR951560-1G
5-Amino-1-ethyl-1H-indazole 98%
5-Amino-1-ethyl-1H-indazole
Vzorec:
C₉H₁₁N₃ MW: 161,204 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16620004 CAS číslo: 511249-17-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...