Tisk…

4-(1,3-Oxazol-5-yl)phenol 98%

Dodavatel: Apollo Scientific

OR951536-1G OR951536-250MG OR951536-5G
APOSOR951536-1GEA 3640 CZK
APOSOR951536-1G APOSOR951536-250MG APOSOR951536-5G
4-(1,3-Oxazol-5-yl)phenol 98%
4-(1,3-Oxazol-5-yl)phenol
Vzorec: C₉H₇NO₂
Storage Temperature: Ambient
MDL Number: MFCD00207683
CAS číslo: 1128-71-8

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR