Tisk…

6-Methoxy-3-pyridineacetic acid 98%

Dodavatel: Apollo Scientific

Synonyms: 6-Methoxy-3-pyridineacetic Acid

OR951528-1G OR951528-5G
APOSOR951528-1GEA 1660 CZK
APOSOR951528-1G APOSOR951528-5G
6-Methoxy-3-pyridineacetic acid 98%
6-Methoxy-3-pyridineacetic acid
Vzorec: C₈H₉NO₃
MW: 167,162 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09924951
CAS číslo: 902130-87-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR