4-(Aminomethyl)-N,N-dimethylbenzamide 98%
Dodavatel: Apollo Scientific
OR951509-1G
OR951509-250MG
APOSOR951509-1GEA
11810
CZK
APOSOR951509-1G
APOSOR951509-250MG
4-(Aminomethyl)-N,N-dimethylbenzamide 98%
4-(Aminomethyl)-N,N-dimethylbenzamide
Vzorec:
C₁₀H₁₄N₂O MW: 178,231 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07186282 CAS číslo: 164648-76-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...