Tisk…

5,6,7,8-Tetrahydroquinazolin-2-amine 95%

Dodavatel: Apollo Scientific

OR951480-1G
APOSOR951480-1GEA 2150 CZK
APOSOR951480-1G
5,6,7,8-Tetrahydroquinazolin-2-amine 95%
5,6,7,8-Tetrahydroquinazolin-2-amine
Vzorec: C₈H₁₁N₃
MW: 149,193 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08060043
CAS číslo: 2305-85-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR