Tisk…

4-(2,5-Dimethyl-pyrrol-1-yl)-phenylamine 98%

Dodavatel: Apollo Scientific

OR951428-10G OR951428-1G OR951428-5G
APOSOR951428-10GEA 27080 CZK
APOSOR951428-10G APOSOR951428-1G APOSOR951428-5G
4-(2,5-Dimethyl-pyrrol-1-yl)-phenylamine 98%
4-(2,5-Dimethyl-pyrrol-1-yl)-phenylamine
Vzorec: C₁₂H₁₄N₂
MW: 186,253 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01125287
CAS číslo: 60176-19-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR