4-(4-Piperidyl)-1-butanol 95%
Dodavatel: Apollo Scientific
OR951389-1G
OR951389-250MG
APOSOR951389-1GEA
6090
CZK
APOSOR951389-1G
APOSOR951389-250MG
4-(4-Piperidyl)-1-butanol 95%
4-(4-Piperidyl)-1-butanol
Vzorec:
C₉H₁₉NO MW: 157,253 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02178381 CAS číslo: 57614-92-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...