Tisk…

2-(Pyrrolidin-1-ylmethyl)benzonitrile 97%

Dodavatel: Apollo Scientific

OR951362-10G OR951362-1G OR951362-5G
APOSOR951362-10GEA 21490 CZK
APOSOR951362-10G APOSOR951362-1G APOSOR951362-5G
2-(Pyrrolidin-1-ylmethyl)benzonitrile 97%
2-(Pyrrolidin-1-ylmethyl)benzonitrile
Vzorec: C₁₂H₁₄N₂
MW: 186,253 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07339548
CAS číslo: 135277-07-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR