Methyl 3-hydroxy-1H-pyrrole-2-carboxylate 95%
Dodavatel: Apollo Scientific
OR951350-1G
OR951350-250MG
APOSOR951350-1GEA
6680
CZK
APOSOR951350-1G
APOSOR951350-250MG
Methyl 3-hydroxy-1H-pyrrole-2-carboxylate 95%
Methyl 3-hydroxy-1H-pyrrole-2-carboxylate
Vzorec:
C₆H₇NO₃ MW: 141,125 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11975599 CAS číslo: 79068-31-8 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...