Methyl 5-formyl-2-methoxybenzoate 98%
Dodavatel: Apollo Scientific
OR951320-100G
OR951320-25G
APOSOR951320-100GEA
6750
CZK
APOSOR951320-100G
APOSOR951320-25G
Methyl 5-formyl-2-methoxybenzoate 98%
Methyl 5-formyl-2-methoxybenzoate
Vzorec:
C₁₀H₁₀O₄ Storage Temperature: Ambient |
MDL Number:
MFCD10565654 CAS číslo: 78515-16-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...