Tisk…

4-Methylpyrazole-3-carboxylic acid 98%

Dodavatel: Apollo Scientific

Synonyms: 4-Methylpyrazole-3-carboxylic Acid

OR951257-5G
APOSOR951257-5GEA 4860 CZK
APOSOR951257-5G
4-Methylpyrazole-3-carboxylic acid 98%
4-Methylpyrazole-3-carboxylic acid
Vzorec: C₅H₆N₂O₂
MW: 126,113 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16495896
CAS číslo: 82231-51-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR