Tisk…

3-Amino-N-cyclopropylbenzamide 96%

Dodavatel: Apollo Scientific

OR951200-1G OR951200-5G
APOSOR951200-1GEA 6620 CZK
APOSOR951200-1G APOSOR951200-5G
3-Amino-N-cyclopropylbenzamide 96%
3-Amino-N-cyclopropylbenzamide
Vzorec: C₁₀H₁₂N₂O
MW: 176,215 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07355824
CAS číslo: 871673-24-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR