5-Isopropyl-1H-indole-2,3-dione 95%
Dodavatel: Apollo Scientific
Synonyms:
5-Isopropylindoline-2,3-dione
OR951185-1G
OR951185-5G
APOSOR951185-1GEA
2440
CZK
APOSOR951185-1G
APOSOR951185-5G
5-Isopropyl-1H-indole-2,3-dione 95%
5-Isopropyl-1H-indole-2,3-dione
Vzorec:
C₁₁H₁₁NO₂ MW: 189,211 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02947184 CAS číslo: 150560-58-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...