Tisk…

4-Pent-1-enyl-benzoic acid 97%

Dodavatel: Apollo Scientific

OR951134-1G OR951134-5G
APOSOR951134-1GEA 6020 CZK
APOSOR951134-1G APOSOR951134-5G
4-Pent-1-enyl-benzoic acid 97%
4-Pent-1-enyl-benzoic acid
Vzorec: C₁₂H₁₄O₂
Storage Temperature: Ambient
MDL Number: MFCD09261414
CAS číslo: 202798-06-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR