4-Methyl-1,2,3,4-tetrahydroisoquinoline hydrochloride 95%
Dodavatel: Apollo Scientific
OR951083-1G
OR951083-250MG
APOSOR951083-1GEA
22950
CZK
APOSOR951083-1G
APOSOR951083-250MG
4-Methyl-1,2,3,4-tetrahydroisoquinoline hydrochloride 95%
4-Methyl-1,2,3,4-tetrahydroisoquinoline hydrochloride
Vzorec:
C₁₀H₁₃N·ClH MW: 183,678 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11501478 CAS číslo: 111661-47-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...